Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CNOT6 Partial ORF (NP_056270.2, 112 a.a. - 210 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$499.00 - $769.00

Specifications

Accession Number NP_056270.2
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 57472
Molecular Weight (g/mol) 36.63kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-940-009
View Documents
Abnova™
H00057472Q01S
10 ug
Each for $499.00
Add to Cart
 
89-940-010
View Documents
Abnova™
H00057472Q01L
25 ug
Each for $769.00
Add to Cart
 
Description

Description

The protein encoded by this gene is a subunit of the CCR4-NOT core transcriptional regulation complex. The encoded protein has a 3'-5' RNase activity and prefers polyadenylated substates. [provided by RefSeq]

Sequence: LPFELGKLFQLQTLGLKGNPLTQDILNLYQEPDGTRRLLNYLLDNLSGTAKRITTEQPPPRSWIMLQEPDRTRPTALFSVMCYNVLCDKYATRQLYGYC
Specifications

Specifications

NP_056270.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CCR4/KIAA1194
CNOT6
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
57472
CNOT6 (Human) Recombinant Protein (Q01)
LPFELGKLFQLQTLGLKGNPLTQDILNLYQEPDGTRRLLNYLLDNLSGTAKRITTEQPPPRSWIMLQEPDRTRPTALFSVMCYNVLCDKYATRQLYGYC
RUO
CNOT6
Wheat Germ (in vitro)
GST
Liquid
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit