Learn More
Novus Biologicals™ Human Collagen III alpha 1/COL3A1 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier: Novus Biologicals™ NBP275858
Description
The Human Collagen III alpha 1/COL3A1 ELISA Kit (Colorimetric) measures human Collagen III alpha 1/COL3A1 in serum, plasma and other biological fluids.Specifications
0.63 - 40 ng/mL | |
This kit recognizes Human COL3a1 in samples. No significant cross-reactivity or interference between Human COL3a1 and analogues was observed. | |
Biological Fluids, Plasma, Serum | |
1281 | |
Human | |
Cancer, Cell Biology, Extracellular Matrix | |
Collagen III alpha 1/COL3A1 |
0.38 ng/mL | |
ELISA Kit (Colorimetric) | |
ELISA | |
COL3A1 | |
1 Kit | |
Storage of components varies. See protocol for specific instructions. |