Learn More
Novus Biologicals™ Human Collagen VI alpha 3 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP275880
Catalog No. NBP275880
Add to cart
Description
Description
The Human Collagen VI alpha 3 ELISA Kit (Colorimetric) measures human Collagen VI alpha 3 in serum, plasma and other biological fluids.
Specifications
Specifications
31.25 - 2000 pg/mL
ELISA
ELISA Kit (Colorimetric)
Storage is content dependent.
Human
1 Kit
This kit recognizes Human COL6a3 in samples. No significant cross-reactivity or interference between Human COL6a3 and analogues was observed.
1293
18.75 pg/mL
Collagen VI alpha 3
COL6A3
Biological Fluids, Plasma, Serum