Learn More
Novus Biologicals™ Human Complement Factor H-related 2/CFHR2/CFHL2 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP275247
Catalog No. NBP275247
Add to cart
Description
Description
The Human Complement Factor H-related 2/CFHR2/CFHL2 ELISA Kit (Colorimetric) measures human Complement Factor H-related 2/CFHR2/CFHL2 in serum, plasma and other biological fluids.
Specifications
Specifications
0.63 - 40 ng/mL
ELISA
ELISA Kit (Colorimetric)
Storage is content dependent.
Human
1 Kit
This kit recognizes Human CFHR2 in samples. No significant cross-reactivity or interference between Human CFHR2 and analogues was observed.
3080
0.38 ng/mL
Complement Factor H-related 2/CFHR2/CFHL2
CFHR2
Biological Fluids, Plasma, Serum