Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Complement Factor H-related 5/ CFHR5 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $503.00
Specifications
Antigen | Complement Factor H-related 5/CFHR5 |
---|---|
Clone | 390513 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunoprecipitation |
Description
Complement Factor H-related 5/CFHR5 Monoclonal specifically detects Complement Factor H-related 5/CFHR5 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Complement Factor H-related 5/CFHR5 | |
LYOPH | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Human | |
Q9BXR6 | |
81494 | |
Mouse myeloma cell line NS0-derived recombinant human Complement Factor H-related 5/CFHR5 Glu19-Glu569 Accession # Q9BXR6 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Complement Factor H-related 5/CFHR5 in direct ELISAs and Western blots. |
390513 | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CFHL5, CFHL5factor H-related protein 5, CFHR5, complement factor H-related 5, Complement Factor Hrelated 5, FHR5, FHR-5complement factor H-related protein 5, FHR5MGC133240, FLJ10549 | |
CFHR5 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title