Learn More
Abnova™ Human COPS6 Full-length ORF (NP_006824.2, 1 a.a. - 327 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00010980P01S
Description
The protein encoded by this gene is one of the eight subunits of COP9 signalosome, a highly conserved protein complex that functions as an important regulator in multiple signaling pathways. The structure and function of COP9 signalosome is similar to that of the 19S regulatory particle of 26S proteasome. COP9 signalosome has been shown to interact with SCF-type E3 ubiquitin ligases and act as a positive regulator of E3 ubiquitin ligases. This protein belongs to translation initiation factor 3 (eIF3) superfamily. It is involved in the regulation of cell cycle and likely to be a cellular cofactor for HIV-1 accessory gene product Vpr. [provided by RefSeq]
Sequence: MAAAAAAAAATNGTGGSSGMEVDAAVVPSVMACGVTGSVSVALHPLVILNISDHWIRMRSQEGRPVQVIGALIGKQEGRNIEVMNSFELLSHTVEEKIIIDKEYYYTKEEQFKQVFKELEFLGWYTTGGPPDPSDIHVHKQVCEIIESPLFLKLNPMTKHTDLPVSVFESVIDIINGEATMLFAELTYTLATEEAERIGVDHVARMTATGSGENSTVAEHLIAQHSAIKMLHSRVKLILEYVKASEAGEVPFNHEILREAYALCHCLPVLSTDKFKTDFYDQCNDVGLMAYLGTITKTCNTMNQFVNKFNVLYDRQGIGRRMRGLFFSpecifications
NP_006824.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
62.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CSN6/MOV34-34KD | |
COPS6 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
10980 | |
COPS6 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAAAAAAAAATNGTGGSSGMEVDAAVVPSVMACGVTGSVSVALHPLVILNISDHWIRMRSQEGRPVQVIGALIGKQEGRNIEVMNSFELLSHTVEEKIIIDKEYYYTKEEQFKQVFKELEFLGWYTTGGPPDPSDIHVHKQVCEIIESPLFLKLNPMTKHTDLPVSVFESVIDIINGEATMLFAELTYTLATEEAERIGVDHVARMTATGSGENSTVAEHLIAQHSAIKMLHSRVKLILEYVKASEAGEVPFNHEILREAYALCHCLPVLSTDKFKTDFYDQCNDVGLMAYLGTITKTCNTMNQFVNKFNVLYDRQGIGRRMRGLFF | |
RUO | |
COPS6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.