Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Corin Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $549.50
Specifications
Antigen | Corin |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, ELISA, Immunohistochemistry 0.3-15 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
Corin Polyclonal specifically detects Corin in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
Corin | |
Western Blot 1 ug/mL, ELISA, Immunohistochemistry 0.3-15 ug/mL | |
Polyclonal | |
Goat | |
Human | |
ATC2, atrial natriuteric peptide-converting enzyme, Corin, corin, serine peptidase, EC 3.4.21, EC 3.4.21.-, heart specific serine proteinase, Heart-specific serine proteinase ATC2, MGC119742, pro-ANP-convertase, Pro-ANP-converting enzyme, PRSC, serine protease, Transmembrane protease serine 10 | |
CORIN | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q9Y5Q5 | |
10699 | |
Mouse myeloma cell line NS0-derived recombinant human Corin Ser67-Asn1042 Accession # Q9Y5Q5 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human Corin in direct ELISAs and Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title