Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CORO1C Full-length ORF (NP_055140.1, 1 a.a. - 474 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Specifications

Name CORO1C (Human) Recombinant Protein (P01)
Recombinant Yes
Purification Method Glutathione Sepharose 4 Fast Flow
For Use With (Application) Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Molecular Weight 79.6kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-940-249
View Documents
Abnova™
H00023603P01L
25 ug This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
89-940-248
View Documents
Abnova™
H00023603P01S
10 ug This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. [provided by RefSeq]

Sequence: MRRVVRQSKFRHVFGQAVKNDQCYDDIRVSRVTWDSSFCAVNPRFVAIIIEASGGGAFLVLPLHKTGRIDKSYPTVCGHTGPVLDIDWCPHNDQVIASGSEDCTVMVWQIPENGLTLSLTEPVVILEGHSKRVGIVAWHPTARNVLLSAGCDNAIIIWNVGTGEALINLDDMHSDMIYNVSWNRNGSLICTASKDKKVRVIDPRKQEIVAEKEKAHEGARPMRAIFLADGNVFTTGFSRMSERQLALWNPKNMQEPIALHEMDTSNGVLLPFYDPDTSIIYLCGKGDSSIRYFEITDESPYVHYLNTFSSKEPQRGMGYMPKRGLDVNKCEIARFFKLHERKCEPIIMTVPRKSDLFQDDLYPDTAGPEAALEAEEWFEGKNADPILISLKHGYIPGKNRDLKVVKKNILDSKPTANKKCDLISIPKKTTDTASVQNEAKLDEILKEIKSIKDTICNQDERISKLEQQMAKIAA
Specifications

Specifications

CORO1C (Human) Recombinant Protein (P01)
Glutathione Sepharose 4 Fast Flow
79.6kDa
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit