Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CPA5 Full-length ORF (NP_525124.2, 1 a.a. - 436 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00093979P01S

 View more versions of this product

Catalog No. 89-940-323


Add to cart

Description

Description

Carboxypeptidases have functions ranging from digestion of food to selective biosynthesis of neuroendocrine peptides. Members of the A/B subfamily of carboxypeptidases, such as CPA5, contain an approximately 90-amino acid pro region that assists in the folding of the active carboxypeptidase domain. Cleavage of the pro region activates the enzyme (Wei et al., 2002 [PubMed 11836249]).[supplied by OMIM]

Sequence: MQGTPGGGTRPGPSPVDRRTLLVFSFILAAALGQMNFTGDQVLRVLAKDEKQLSLLGDLEGLKPQKVDFWRGPARPSLPVDMRVPFSELKDIKAYLESHGLAYSIMIKDIQVLLDEERQAMAKSRRLERSTNSFSYSSYHTLEEIYSWIDNFVMEHSDIVSKIQIGNSFENQSILVLKFSTGGSRHPAIWIDTGIHSREWITHATGIWTANKIVSDYGKDRVLTDILNAMDIFIELVTNPDGFAFTHSMNRLWRKNKSIRPGIFCIGVDLNRNWKSGFGGNGSNSNPCSETYHGPSPQSEPEVAAIVNFITAHGNFKALISIHSYSQMLMYPYGRSLDPVSNQRELYDLAKDAVEALYKVHGIEYIFGSISTTLYVASGITVDWAYDSGIKYAFSFELRDTGQYGFLLPATQIIPTAQETWMALRTIMEHTLNHPY
Specifications

Specifications

NP_525124.2
Liquid
93979
CPA5 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MQGTPGGGTRPGPSPVDRRTLLVFSFILAAALGQMNFTGDQVLRVLAKDEKQLSLLGDLEGLKPQKVDFWRGPARPSLPVDMRVPFSELKDIKAYLESHGLAYSIMIKDIQVLLDEERQAMAKSRRLERSTNSFSYSSYHTLEEIYSWIDNFVMEHSDIVSKIQIGNSFENQSILVLKFSTGGSRHPAIWIDTGIHSREWITHATGIWTANKIVSDYGKDRVLTDILNAMDIFIELVTNPDGFAFTHSMNRLWRKNKSIRPGIFCIGVDLNRNWKSGFGGNGSNSNPCSETYHGPSPQSEPEVAAIVNFITAHGNFKALISIHSYSQMLMYPYGRSLDPVSNQRELYDLAKDAVEALYKVHGIEYIFGSISTTLYVASGITVDWAYDSGIKYAFSFELRDTGQYGFLLPATQIIPTAQETWMALRTIMEHTLNHPY
RUO
CPA5
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
75.4kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ35969
CPA5
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit