Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CPNE2 Full-length ORF (NP_689940.3, 1 a.a. - 548 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00221184P01

Catalog No. 89-940-357


Add to cart

Description

Description

Calcium-dependent membrane-binding proteins may regulate molecular events at the interface of the cell membrane and cytoplasm. This gene is one of several genes that encode a calcium-dependent protein containing two N-terminal type II C2 domains and an integrin A domain-like sequence in the C-terminus. Sequence analysis identified multiple alternatively spliced transcript variants but their full-length natures could not be determined. [provided by RefSeq]

Sequence: MAHIPSGGAPAAGAAPMGPQYCVCKVELSVSGQNLLDRDVTSKSDPFCVLFTENNGRWIEYDRTETAINNLNPAFSKKFVLDYHFEEVQKLKFALFDQDKSSMRLDEHDFLGQFSCSLGTIVSSKKITRPLLLLNDKPAGKGLITIAAQELSDNRVITLSLAGRRLDKKDLFGKSDPFLEFYKPGDDGKWMLVHRTEVIKYTLDPVWKPFTVPLVSLCDGDMEKPIQVMCYDYDNDGGHDFIGEFQTSVSQMCEARDSVPLEFECINPKKQRKKKNYKNSGIIILRSCKINRDYSFLDYILGGCQLMFTVGIDFTASNGNPLDPSSLHYINPMGTNEYLSAIWAVGQIIQDYDSDKMFPALGFGAQLPPDWKVSHEFAINFNPTNPFCSGVDGIAQAYSACLPHIRFYGPTNFSPIVNHVARFAAQATQQRTATQYFILLIITDGVISDMEETRHAVVQASKLPMSIIIVGVGNADFAAMEFLDGDSRMLRSHTGEEAARDIVQFVPFREFRNAAKETLAKAVLAELPQQVVQYFKHKNLPPTNSEPA
Specifications

Specifications

NP_689940.3
Liquid
221184
CPNE2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAHIPSGGAPAAGAAPMGPQYCVCKVELSVSGQNLLDRDVTSKSDPFCVLFTENNGRWIEYDRTETAINNLNPAFSKKFVLDYHFEEVQKLKFALFDQDKSSMRLDEHDFLGQFSCSLGTIVSSKKITRPLLLLNDKPAGKGLITIAAQELSDNRVITLSLAGRRLDKKDLFGKSDPFLEFYKPGDDGKWMLVHRTEVIKYTLDPVWKPFTVPLVSLCDGDMEKPIQVMCYDYDNDGGHDFIGEFQTSVSQMCEARDSVPLEFECINPKKQRKKKNYKNSGIIILRSCKINRDYSFLDYILGGCQLMFTVGIDFTASNGNPLDPSSLHYINPMGTNEYLSAIWAVGQIIQDYDSDKMFPALGFGAQLPPDWKVSHEFAINFNPTNPFCSGVDGIAQAYSACLPHIRFYGPTNFSPIVNHVARFAAQATQQRTATQYFILLIITDGVISDMEETRHAVVQASKLPMSIIIVGVGNADFAAMEFLDGDSRMLRSHTGEEAARDIVQFVPFREFRNAAKETLAKAVLAELPQQVVQYFKHKNLPPTNSEPA
RUO
CPNE2
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
87.6kDa
Glutathione Sepharose 4 Fast Flow
2 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
COPN2|CPN2|DKFZp686E06199|MGC16924
CPNE2
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit