Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CPNE7 Partial ORF (NP_705900, 460 a.a. - 558 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00027132Q01S

 View more versions of this product

Catalog No. 89-940-371


Add to cart

Description

Description

This gene encodes a member of the copine family, which is composed of calcium-dependent membrane-binding proteins. The gene product contains two N-terminal C2 domains and one von Willebrand factor A domain. The encoded protein may be involved in membrane trafficking. Two alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: ASRLPMSIIIVGVGNADFTDMQVLDGDDGVLRSPRGEPALRDIVQFVPFRELKNASPAALAKCVLAEVPKQVVEYYSHRGLPPRSLGVPAGEASPGCTP
Specifications

Specifications

NP_705900
Liquid
27132
CPNE7 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC34192
CPNE7
Wheat Germ (in vitro)
GST
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
ASRLPMSIIIVGVGNADFTDMQVLDGDDGVLRSPRGEPALRDIVQFVPFRELKNASPAALAKCVLAEVPKQVVEYYSHRGLPPRSLGVPAGEASPGCTP
RUO
CPNE7
wheat germ expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit