Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CPOX Full-length ORF (AAH23554.1, 1 a.a. - 454 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00001371P01

Catalog No. 89-940-375


Add to cart

Description

Description

Coproporphyrinogen oxidase (EC 1.3.3.3) is the sixth enzyme of the heme biosynthetic pathway. This soluble protein is localized in the intermembrane space of mitochondria and catalyzes the stepwise oxidative decarboxylation of the heme precursor coproporphyrinogen III to protoporphyrinogen IX via the tricarboxylic intermediate harderoporphyrinogen.[supplied by OMIM]

Sequence: MALQLGRLSSGPCWLVARGGCGGPRAWSQCGGGGLRAWSQRSAAGRVCRPPGPAGTEQSRGLGHGSTSRGGPWVGTGLAAALAGLVGLATAAFGHVQRAEMLPKTSGTRATSLGRPEEEEDELAHRCSSFMAPPVTDLGELRRRPGDMKTKMELLILETQAQVCQALAQVDGGANFSVDRWERKEGGGGISCVLQDGCVFEKAGVSISVVHGNLSEEAAKQMRSRGKVLKTKDGKLPFCAMGVSSVIHPKNPHAPTIHFNYRYFEVEEADGNKQWWFGGGCDLTPTYLNQEDAIHFHRTLKEACDQHGPDLYPKFKKWCDDYFFIAHRGERRGIGGIFFDDLDSPSKEEVFRFVQSCARAVVPSYIPLVKKHCDDSFTPQEKLWQQLRRGRYVEFNLLYDRGTKFGLFTPGSRIESILMSLPLTARWEYMHSPSENSKEAEILEVLRHPRDWVR
Specifications

Specifications

AAH23554.1
Liquid
1371
CPOX (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MALQLGRLSSGPCWLVARGGCGGPRAWSQCGGGGLRAWSQRSAAGRVCRPPGPAGTEQSRGLGHGSTSRGGPWVGTGLAAALAGLVGLATAAFGHVQRAEMLPKTSGTRATSLGRPEEEEDELAHRCSSFMAPPVTDLGELRRRPGDMKTKMELLILETQAQVCQALAQVDGGANFSVDRWERKEGGGGISCVLQDGCVFEKAGVSISVVHGNLSEEAAKQMRSRGKVLKTKDGKLPFCAMGVSSVIHPKNPHAPTIHFNYRYFEVEEADGNKQWWFGGGCDLTPTYLNQEDAIHFHRTLKEACDQHGPDLYPKFKKWCDDYFFIAHRGERRGIGGIFFDDLDSPSKEEVFRFVQSCARAVVPSYIPLVKKHCDDSFTPQEKLWQQLRRGRYVEFNLLYDRGTKFGLFTPGSRIESILMSLPLTARWEYMHSPSENSKEAEILEVLRHPRDWVR
RUO
CPOX
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
76.6kDa
Glutathione Sepharose 4 Fast Flow
2 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CPO|CPX|HCP
CPOX
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit