Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CPSF6 Full-length ORF (AAH05000.1, 1 a.a. - 478 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00011052P01L

 View more versions of this product

Catalog No. 89-940-397


Add to cart

Description

Description

The protein encoded by this gene is one subunit of a cleavage factor required for 3' RNA cleavage and polyadenylation processing. The interaction of the protein with the RNA is one of the earliest steps in the assembly of the 3' end processing complex and facilitates the recruitment of other processing factors. The cleavage factor complex is composed of four polypeptides. This gene encodes the 68kD subunit. It has a domain organization reminiscent of spliceosomal proteins. [provided by RefSeq]

Sequence: MADGVDHIDIYADVGEEFNQEAEYGGHDQIDLYDDVISPSANNGDAPEDRDYMDTLPPTVGDDVGKGAAPNVVYTYTGKRIALYIGNLTWWTTDEDLTEAVHSLGVNDILEIKFFENRANGQSKGFALVGVGSEASSKKLMDLLPKRELHGQNPVVTPCNKQFLSQFEMQSRKTTQSGQMSGEGKAGPPLAGPPNRGDRPPPPVLFPGQPFGQPPLGPLPPGPPPPVPGYGPPPGPPPPQQGPPPPPGPFPPRPPGPLGPPLTLAPPPHLPGPPPGAPPPAPHVNPAFFPPPTNSGMPTSDSRGPPPTDPYGRPPPYDRGDYGPPGREMDTARTPLSEAEFEEIMNRNRAISSSAISRAVSDASAGDYGSAIETLVTAISLIKQSKVSADDRCKVLISSLQDCLHGIESKSYGSGSRRERSRERDHSRSREKSRRHKSRSRDRHDDYYRERSRERERHRDRDRDRDRERDREREYRHR
Specifications

Specifications

AAH05000.1
Liquid
11052
CPSF6 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MADGVDHIDIYADVGEEFNQEAEYGGHDQIDLYDDVISPSANNGDAPEDRDYMDTLPPTVGDDVGKGAAPNVVYTYTGKRIALYIGNLTWWTTDEDLTEAVHSLGVNDILEIKFFENRANGQSKGFALVGVGSEASSKKLMDLLPKRELHGQNPVVTPCNKQFLSQFEMQSRKTTQSGQMSGEGKAGPPLAGPPNRGDRPPPPVLFPGQPFGQPPLGPLPPGPPPPVPGYGPPPGPPPPQQGPPPPPGPFPPRPPGPLGPPLTLAPPPHLPGPPPGAPPPAPHVNPAFFPPPTNSGMPTSDSRGPPPTDPYGRPPPYDRGDYGPPGREMDTARTPLSEAEFEEIMNRNRAISSSAISRAVSDASAGDYGSAIETLVTAISLIKQSKVSADDRCKVLISSLQDCLHGIESKSYGSGSRRERSRERDHSRSREKSRRHKSRSRDRHDDYYRERSRERERHRDRDRDRDRERDREREYRHR
RUO
CPSF6
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
78.7kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CFIM|CFIM68|HPBRII-4|HPBRII-7
CPSF6
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit