Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CRAT Full-length ORF (NP_000746.2, 1 a.a. - 626 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00001384P01

Catalog No. 89-940-434


Add to cart

Description

Description

This gene encodes carnitine acetyltransferase (CRAT), which is a key enzyme in the metabolic pathway in mitochondria, peroxisomes and endoplasmic reticulum. CRAT catalyzes the reversible transfer of acyl groups from an acyl-CoA thioester to carnitine and regulates the ratio of acylCoA/CoA in the subcellular compartments. Alternate splicing results in multiple transcript variants; additional transcript variants have been described, but their biological validity has not been determined. [provided by RefSeq]

Sequence: MLAFAARTVVKPLGFLKPFSLMKASSRFKAHQDALPRLPVPPLQQSLDHYLKALQPIVSEEEWAHTKQLVDEFQASGGVGERLQKGLERRARKTENWLSEWWLKTAYLQYRQPVVIYSSPGVMLPKQDFVDLQGQLRFAAKLIEGVLDFKVMIDNETLPVEYLGGKPLCMNQYYQILSSCRVPGPKQDTVSNFSKTKKPPTHITVVHNYQFFELDVYHSDGTPLTADQIFVQLEKIWNSSLQTNKEPVGILTSNHRNSWAKAYNTLIKDKVNRDSVRSIQKSIFTVCLDATMPRVSEDVYRSHVAGQMLHGGGSRLNSGNRWFDKTLQFIVAEDGSCGLVYEHAAAEGPPIVTLLDYVIEYTKKPELVRSPMVPLPMPKKLRFNITPEIKSDIEKAKQNLSIMIQDLDITVMVFHHFGKDFPKSEKLSPDAFIQMALQLAYYRIYGQACATYESASLRMFHLGRTDTIRSASMDSLTFVKAMDDSSVTEHQKVELLRKAVQAHRGYTDRAIRGEAFDRHLLGLKLQAIEDLVSMPDIFMDTSYAIAMHFHLSTSQVPAKTDCVMFFGPVVPDGYGVCYNPMEAHINFSLSAYNSCAETNAARLAHYLEKALLDMRALLQSHPRAKL
Specifications

Specifications

NP_000746.2
Liquid
1384
CRAT (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MLAFAARTVVKPLGFLKPFSLMKASSRFKAHQDALPRLPVPPLQQSLDHYLKALQPIVSEEEWAHTKQLVDEFQASGGVGERLQKGLERRARKTENWLSEWWLKTAYLQYRQPVVIYSSPGVMLPKQDFVDLQGQLRFAAKLIEGVLDFKVMIDNETLPVEYLGGKPLCMNQYYQILSSCRVPGPKQDTVSNFSKTKKPPTHITVVHNYQFFELDVYHSDGTPLTADQIFVQLEKIWNSSLQTNKEPVGILTSNHRNSWAKAYNTLIKDKVNRDSVRSIQKSIFTVCLDATMPRVSEDVYRSHVAGQMLHGGGSRLNSGNRWFDKTLQFIVAEDGSCGLVYEHAAAEGPPIVTLLDYVIEYTKKPELVRSPMVPLPMPKKLRFNITPEIKSDIEKAKQNLSIMIQDLDITVMVFHHFGKDFPKSEKLSPDAFIQMALQLAYYRIYGQACATYESASLRMFHLGRTDTIRSASMDSLTFVKAMDDSSVTEHQKVELLRKAVQAHRGYTDRAIRGEAFDRHLLGLKLQAIEDLVSMPDIFMDTSYAIAMHFHLSTSQVPAKTDCVMFFGPVVPDGYGVCYNPMEAHINFSLSAYNSCAETNAARLAHYLEKALLDMRALLQSHPRAKL
RUO
CRAT
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
97.3kDa
Glutathione Sepharose 4 Fast Flow
2 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CAT1
CRAT
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit