Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CRAT Partial ORF (NP_659006, 445 a.a. - 544 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00001384Q01S

 View more versions of this product

Catalog No. 89-940-432


Add to cart

Description

Description

This gene encodes carnitine acetyltransferase (CRAT), which is a key enzyme in the metabolic pathway in mitochondria, peroxisomes and endoplasmic reticulum. CRAT catalyzes the reversible transfer of acyl groups from an acyl-CoA thioester to carnitine and regulates the ratio of acylCoA/CoA in the subcellular compartments. Alternate splicing results in multiple transcript variants; additional transcript variants have been described, but their biological validity has not been determined. [provided by RefSeq]

Sequence: AIEDLVSMPDIFMDTSYAIAMHFHLSTSQVPAKTDCVMFFGPVVPDGYGVCYNPMEAHINFSLSAYNSCAETNAARLAHYLEKALLDMRALLQSHPRAKL
Specifications

Specifications

NP_659006
Liquid
1384
CRAT (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CAT1
CRAT
Wheat Germ (in vitro)
GST
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
AIEDLVSMPDIFMDTSYAIAMHFHLSTSQVPAKTDCVMFFGPVVPDGYGVCYNPMEAHINFSLSAYNSCAETNAARLAHYLEKALLDMRALLQSHPRAKL
RUO
CRAT
wheat germ expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit