Learn More
Abnova™ Human CRB1 Partial ORF (NP_036208, 26 a.a. - 134 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$499.00 - $769.00
Specifications
Accession Number | NP_036208 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 23418 |
Molecular Weight (g/mol) | 37.73kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-940-436
|
Abnova™
H00023418Q01L |
25 ug |
Each for $769.00
|
|
|||||
89-940-435
|
Abnova™
H00023418Q01S |
10 ug |
Each for $499.00
|
|
|||||
Description
This gene encodes a protein which is similar to the Drosophila crumbs protein and localizes to the inner segment of mammalian photoreceptors. In Drosophila crumbs localizes to the stalk of the fly photoreceptor and may be a component of the molecular scaffold that controls proper development of polarity in the eye. Mutations in this gene are associated with a severe form of retinitis pigmentosa, RP12, and with Leber congenital amaurosis. Alternatively spliced transcript variants have been observed but their full-length nature has yet to be determined. [provided by RefSeq]
Sequence: FCNKNNTRCLSNSCQNNSTCKDFSKDNDCSCSDTANNLDKDCDNMKDPCFSNPCQGSATCVNTPGERSFLCKCPPGYSGTICETTIGSCGKNSCQHGGICHQDPIYPVCSpecifications
NP_036208 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
LCA8/RP12 | |
CRB1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
23418 | |
CRB1 (Human) Recombinant Protein (Q01) | |
FCNKNNTRCLSNSCQNNSTCKDFSKDNDCSCSDTANNLDKDCDNMKDPCFSNPCQGSATCVNTPGERSFLCKCPPGYSGTICETTIGSCGKNSCQHGGICHQDPIYPVC | |
RUO | |
CRB1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.