Learn More
Novus Biologicals™ Human Creatine Kinase, Muscle/CKMM ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP275305
Catalog No. NBP275305
Add to cart
Description
Description
The Human Creatine Kinase, Muscle/CKMM ELISA Kit (Colorimetric) measures human Creatine Kinase, Muscle/CKMM in serum, plasma and other biological fluids.
Specifications
Specifications
1.56 - 100 ng/mL
ELISA
ELISA Kit (Colorimetric)
Storage is content dependent.
Human
1 Kit
This kit recognizes Human CKM in samples. No significant cross-reactivity or interference between Human CKM and analogues was observed.
1158
0.94 ng/mL
Creatine Kinase, Muscle/CKMM
CKM
Biological Fluids, Plasma, Serum