Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CREB3L2 Partial ORF (NP_919047.2, 421 a.a. - 518 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00064764Q01L

 View more versions of this product

Catalog No. 89-940-463


Add to cart

Description

Description

CREB3L2 is a member of the old astrocyte specifically induced substance (OASIS) DNA binding and basic leucine zipper dimerization (bZIP) family of transcription factors, which includes CREB3 (MIM 606443) and CREB4 (MIM 607138).[supplied by OMIM]

Sequence: ASVVRSRNLLIYEEHSPPEESSSPGSAGELGGWDRGSSLLRVSGLESRPDVDLPHFIISNETSLEKSVLLELQQHLVSAKLEGNETLKVVELDRRVNT
Specifications

Specifications

NP_919047.2
Liquid
64764
CREB3L2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BBF2H7|MGC131709|MGC71006
CREB3L2
Wheat Germ (in vitro)
GST
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.52kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
ASVVRSRNLLIYEEHSPPEESSSPGSAGELGGWDRGSSLLRVSGLESRPDVDLPHFIISNETSLEKSVLLELQQHLVSAKLEGNETLKVVELDRRVNT
RUO
CREB3L2
wheat germ expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit