Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CREB3L4 Partial ORF (NP_570968.1, 324 a.a. - 395 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00148327Q02L

 View more versions of this product

Catalog No. 89-940-469


Add to cart

Description

Description

cAMP response element-binding (CREB) proteins are a family of mammalian transcription activators. For further background information on CREB proteins, see CREB1 (MIM 123810).[supplied by OMIM]

Sequence: SEDYQPHGVTSRNILTHKDVTENLETQVVESRLREPPGAKDANGSTRTLLEKMGGKPRPSGRIRSVLHADEM
Specifications

Specifications

NP_570968.1
Liquid
148327
CREB3L4 (Human) Recombinant Protein (Q02)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AIBZIP|ATCE1|CREB3|CREB4|JAL|hJAL
CREB3L4
Wheat Germ (in vitro)
GST
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
33.66kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
SEDYQPHGVTSRNILTHKDVTENLETQVVESRLREPPGAKDANGSTRTLLEKMGGKPRPSGRIRSVLHADEM
RUO
CREB3L4
wheat germ expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit