Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CREG1 (NP_003842, 32 a.a. - 220 a.a.) Partial Recombinant Protein with His tag

Used for SDS-PAGE

Manufacturer:  Abnova™ P5861

Catalog No. 89-940-486


Add to cart

Description

Description

The adenovirus E1A protein both activates and represses gene expression to promote cellular proliferation and inhibit differentiation. The protein encoded by this gene antagonizes transcriptional activation and cellular transformation by E1A. This protein shares limited sequence similarity with E1A and binds both the general transcription factor TBP and the tumor suppressor pRb in vitro. This gene may contribute to the transcriptional control of cell growth and differentiation. [provided by RefSeq]

Sequence: MGSSHHHHHHSSGLVPRGSHMGSMRGGRDHGDWDEASRLPPLPPREDAARVARFVTHVSDWGALATISTLEAVRGRPFADVLSLSDGPPGAGSGVPYFYLSPLQLSVSNLQENPYATLTMTLAQTNFCKKHGFDPQSPLCVHIMLSGTVTKVNETEMDIAKHSLFIRHPEMKTWPSSHNWFFAKLNITNIWVLDYFGGPKIVTPEEYYNVTVQ
Specifications

Specifications

Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
23.6kDa
Escherichia coli expression system
3 ug/lane in 15% SDS-PAGE Stained with Coomassie Blue. Due to the protein nature, dimmers and multimers may be observed.
MGSSHHHHHHSSGLVPRGSHMGSMRGGRDHGDWDEASRLPPLPPREDAARVARFVTHVSDWGALATISTLEAVRGRPFADVLSLSDGPPGAGSGVPYFYLSPLQLSVSNLQENPYATLTMTLAQTNFCKKHGFDPQSPLCVHIMLSGTVTKVNETEMDIAKHSLFIRHPEMKTWPSSHNWFFAKLNITNIWVLDYFGGPKIVTPEEYYNVTVQ
RUO
CREG1
Escherichia coli expression system
Yes
Liquid
8804
CREG1 (Human) Recombinant Protein
>95% by SDS - PAGE
100 ug
Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing.
CREG
CREG1
Escherichia coli
His
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit