Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

Abnova™ Human CRIM1 Partial ORF (NP_057525, 36 a.a. - 145 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00051232Q01L

 View more versions of this product

Catalog No. 89-940-517



Description

Description

Motor neurons are among the earliest neurons to appear after the commencement of cell patterning and the beginning of cell differentiation. Differentiation occurs in a ventral-to-dorsal gradient and is mediated, at least in part, by the concentration of ventrally expressed Sonic hedgehog protein (SHH; MIM 600725). Dorsally expressed factors, such as members of the bone morphogenic protein (e.g., BMP4; MIM 112262) and transforming growth factor-beta (e.g., TGFB1; MIM 190180) families, can repress the induction of these neurons. CRIM1 may interact with growth factors implicated in motor neuron differentiation and survival (Kolle et al., 2000 [PubMed 10642437]).[supplied by OMIM]

Sequence: VCLPCDESKCEEPRNCPGSIVQGVCGCCYTCASQRNESCGGTFGIYGTCDRGLRCVIRPPLNGDSLTEYEAGVCEDENWTDDQLLGFKPCNENLIAGCNIINGKCECNTI
Specifications

Specifications

NP_057525
Liquid
51232
CRIM1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC138194|S52
CRIM1
Wheat Germ (in vitro)
GST
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
VCLPCDESKCEEPRNCPGSIVQGVCGCCYTCASQRNESCGGTFGIYGTCDRGLRCVIRPPLNGDSLTEYEAGVCEDENWTDDQLLGFKPCNENLIAGCNIINGKCECNTI
RUO
CRIM1
wheat germ expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit