Learn More
Human Cripto (aa 31-172) His Tag, Recombinant Protein, Invitrogen™
HEK293 cells Recombinant Protein
Manufacturer:  Thermo Scientific RP87818
Encompass
 View more versions of this product
Catalog No. PIRP87818
Add to cart
Description
Description
Cripto (TDGF1) is a GPI-anchored cell membrane proetin involved in Nodal signaling. Cell associated TDGF1 acts as a a Nodeal coreceptor in cis. Shedding of TDGF1 by TMEM8A modulated Nodal signaling by allowing soluble TDGF1 to act as a Nodal coreceptor on other cells. TDGF1 is expressed both in ES cells and during the early phases of embryo development, while in adults it is reactivated in a wide range of epithelial cancers.
Specifications
Specifications
P13385
Control, ELISA, Western Blot
Dulbecco's PBS with no preservative; pH 7.2
17.4 kD
>95%
HEK293 cells
RUO
TDGF1
Unlabeled
200 µg/mL
Liquid
6997
Human Cripto (aa 31-172) His Tag
25 µg
-20°C
CR; CR1; CRGF; CRIPTO; cripto growth factor; Cripto-1; cripto-1 growth factor; Cripto-3; epidermal growth factor-like Cripto protein; Epidermal growth factor-like cripto protein CR1; Tdgf1; TDGF2; teratocarcinoma-derived growth factor; teratocarcinoma-derived growth factor 1; TERATOCARCINOMA-DERIVED GROWTH FACTOR PRECURSOR (EPIDERMAL GROWTH FACTOR-LIKE CRIPTO PROTEIN) (CRIPTO GROWTH FACTOR)
Human
Yes