Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Cripto (aa 31-172) His Tag, Recombinant Protein, Invitrogen™ Non-distribution product as customer accommodation.

HEK293 cells Recombinant Protein

$147.70 - $263.50

Specifications

Recombinant Yes
Name Human Cripto (aa 31-172) His Tag
For Use With (Application) Control, ELISA, Western Blot
Concentration 200 µg/mL
Conjugate Unlabeled
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIRP87817 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87817
10 µg
Each for $147.70
Add to cart
 
PIRP87818 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87818
25 µg
Each for $263.50
Add to cart
 
Description

Description

Cripto (TDGF1) is a GPI-anchored cell membrane proetin involved in Nodal signaling. Cell associated TDGF1 acts as a a Nodeal coreceptor in cis. Shedding of TDGF1 by TMEM8A modulated Nodal signaling by allowing soluble TDGF1 to act as a Nodal coreceptor on other cells. TDGF1 is expressed both in ES cells and during the early phases of embryo development, while in adults it is reactivated in a wide range of epithelial cancers.
Specifications

Specifications

Yes
Control, ELISA, Western Blot
Unlabeled
Dulbecco's PBS with no preservative; pH 7.2
HEK293 cells
-20°C
P13385
TDGF1
>95%
Human Cripto (aa 31-172) His Tag
200 µg/mL
Liquid
CR; CR1; CRGF; CRIPTO; cripto growth factor; Cripto-1; cripto-1 growth factor; Cripto-3; epidermal growth factor-like Cripto protein; Epidermal growth factor-like cripto protein CR1; Tdgf1; TDGF2; teratocarcinoma-derived growth factor; teratocarcinoma-derived growth factor 1; TERATOCARCINOMA-DERIVED GROWTH FACTOR PRECURSOR (EPIDERMAL GROWTH FACTOR-LIKE CRIPTO PROTEIN) (CRIPTO GROWTH FACTOR)
RUO
Human
6997
17.4 kD
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit