Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CRISP1 Full-length ORF (NP_001122.2, 1 a.a. - 249 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00000167P01L

 View more versions of this product

Catalog No. 89-940-531


Add to cart

Description

Description

Fertilization consists of a sequence of specific cell-cell interactions culminating in the fusion of the sperm and egg plasma membranes. Recognition, binding, and fusion occur through the interaction of complementary molecules that are localized to specific domains of the sperm and egg plasma membranes. In the sperm, the postacrosomal region or equatorial segment is involved in sperm-egg plasma membrane fusion. The protein encoded by this gene is a member of the cysteine-rich secretory protein (CRISP) family. This protein is expressed in the epididymis, is secreted into the epididymal lumen, and binds to the postacrosomal region of the sperm head where it plays a role at fertilization in sperm-egg fusion through complementary sites localized on the egg surface. Two isoforms are encoded by transcript variants of this gene. [provided by RefSeq]

Sequence: MEIKHLLFLVAAACLLPMLSMKKKSARDQFNKLVTDLPNVQEEIVNIHNALRRRVVPPASNMLKMSWSEEAAQNARIFSKYCDMTESNPLERRLPNTFCGENMHMTSYPVSWSSVIGVWYSESTSFKHGEWTTTDDDITTDHYTQIVWATSYLIGCAIASCRQQGSPRYLYVCHYCHEGNDPETKNEPYKTGVPCEACPSNCEDKLCTNPCIYYDEYFDCDIQVHYLGCNHSTTILFCKATCLCDTEIK
Specifications

Specifications

NP_001122.2
Liquid
167
CRISP1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MEIKHLLFLVAAACLLPMLSMKKKSARDQFNKLVTDLPNVQEEIVNIHNALRRRVVPPASNMLKMSWSEEAAQNARIFSKYCDMTESNPLERRLPNTFCGENMHMTSYPVSWSSVIGVWYSESTSFKHGEWTTTDDDITTDHYTQIVWATSYLIGCAIASCRQQGSPRYLYVCHYCHEGNDPETKNEPYKTGVPCEACPSNCEDKLCTNPCIYYDEYFDCDIQVHYLGCNHSTTILFCKATCLCDTEIK
RUO
CRISP1
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
54.9kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AEGL1|ARP|CRISP-1|HSCRISP1D|HSCRISP1G|HUMARP
CRISP1
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit