Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CRNN Full-length ORF (NP_057274.1, 1 a.a. - 495 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00049860P02L

 View more versions of this product

Catalog No. 89-940-565


Add to cart

Description

Description

This gene encodes a member of the “fused gene” family of proteins, which contain N-terminus EF-hand domains and multiple tandem peptide repeats. The encoded protein contains two EF-hand Ca2+ binding domains in its N-terminus and two glutamine- and threonine-rich 60 amino acid repeats in its C-terminus. This gene, also known as squamous epithelial heat shock protein 53, may play a role in the mucosal/epithelial immune response and epidermal differentiation. [provided by RefSeq]

Sequence: MPQLLQNINGIIEAFRRYARTEGNCTALTRGELKRLLEQEFADVIVKPHDPATVDEVLRLLDEDHTGTVEFKEFLVLVFKVAQACFKTLSESAEGACGSQESGSLHSGASQELGEGQRSGTEVGRAGKGQHYEGSSHRQSQQGSRGQNRPGVQTQGQATGSAWVSSYDRQAESQSQERISPQIQLSGQTEQTQKAGEGKRNQTTEMRPERQPQTREQDRAHQTGETVTGSGTQTQAGATQTVEQDSSHQTGRTSKQTQEATNDQNRGTETHGQGRSQTSQAVTGGHAQIQAGTHTQTPTQTVEQDSSHQTGSTSTQTQESTNGQNRGTEIHGQGRSQTSQAVTGGHTQIQAGSHTETVEQDRSQTVSHGGAREQGQTQTQPGSGQRWMQVSNPEAGETVPGGQAQTGASTESGRQEWSSTHPRRCVTEGQGDRQPTVVGEEWVDDHSRETVILRLDQGNLHTSVSSAQGQDAAQSEEKRGITARELYSYLRSTKP
Specifications

Specifications

NP_057274.1
Liquid
49860
CRNN (Human) Recombinant Protein (P02)
12.5% SDS-PAGE Stained with Coomassie Blue.
MPQLLQNINGIIEAFRRYARTEGNCTALTRGELKRLLEQEFADVIVKPHDPATVDEVLRLLDEDHTGTVEFKEFLVLVFKVAQACFKTLSESAEGACGSQESGSLHSGASQELGEGQRSGTEVGRAGKGQHYEGSSHRQSQQGSRGQNRPGVQTQGQATGSAWVSSYDRQAESQSQERISPQIQLSGQTEQTQKAGEGKRNQTTEMRPERQPQTREQDRAHQTGETVTGSGTQTQAGATQTVEQDSSHQTGRTSKQTQEATNDQNRGTETHGQGRSQTSQAVTGGHAQIQAGTHTQTPTQTVEQDSSHQTGSTSTQTQESTNGQNRGTEIHGQGRSQTSQAVTGGHTQIQAGSHTETVEQDRSQTVSHGGAREQGQTQTQPGSGQRWMQVSNPEAGETVPGGQAQTGASTESGRQEWSSTHPRRCVTEGQGDRQPTVVGEEWVDDHSRETVILRLDQGNLHTSVSSAQGQDAAQSEEKRGITARELYSYLRSTKP
RUO
CRNN
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
79.9kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C1orf10|DRC1|PDRC1|SEP53
CRNN
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit