Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CROT Full-length ORF (NP_066974.2, 1 a.a. - 612 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00054677P01S

 View more versions of this product

Catalog No. 89-940-572


Add to cart

Description

Description

This gene encodes a member of the carnitine/choline acetyltransferase family. The encoded protein converts 4,8-dimethylnonanoyl-CoA to its corresponding carnitine ester. This transesterification occurs in the peroxisome and is necessary for transport of medium- and long- chain acyl-CoA molecules out of the peroxisome to the cytosol and mitochondria. The protein thus plays a role in lipid metabolism and fatty acid beta-oxidation. Alternatively spliced transcript variants have been described

Sequence: MENQLAKSTEERTFQYQDSLPSLPVPSLEESLKKYLESVKPFANQEEYKKTEEIVQKFQSGIGEKLHQKLLERAKGKRNWLEEWWLNVAYLDVRIPSQLNVNFAGPAAHFEHYWPPKEGTQLERGSITLWHNLNYWQLLRKEKVPVHKVGNTPLDMNQFRMLFSTCKVPGITRDSIMNYFRTESEGRSPNHIVVLCRGRAFVFDVIHEGCLVTPPELLRQLTYIHKKCHSEPDGPGIAALTSEERTRWAKAREYLIGLDPENLALLEKIQSSLLVYSMEDSSPHVTPEDYSEIIAAILIGDPTVRWGDKSYNLISFSNGVFGCNCDHAPFDAMIMVNISYYVDEKIFQNEGRWKGSEKVRDIPLPEELIFIVDEKVLNDINQAKAQYLREASDLQIAAYAFTSFGKKLTKNKMLHPDTFIQLALQLAYYRLHGHPGCCYETAMTRHFYHGRTETMRSCTVEAVRWCQSMQDPSVNLRERQQKMLQAFAKHNKMMKDCSAGKGFDRHLLGLLLIAKEEGLPVPELFTDPLFSKSGGGGNFVLSTSLVGYLRVQGVVVPMVHNGYGFFYHIRDDRFVVACSAWKSCPETDAEKLVQLTFCAFHDMIQLMNSTHL
Specifications

Specifications

NP_066974.2
Liquid
54677
CROT (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MENQLAKSTEERTFQYQDSLPSLPVPSLEESLKKYLESVKPFANQEEYKKTEEIVQKFQSGIGEKLHQKLLERAKGKRNWLEEWWLNVAYLDVRIPSQLNVNFAGPAAHFEHYWPPKEGTQLERGSITLWHNLNYWQLLRKEKVPVHKVGNTPLDMNQFRMLFSTCKVPGITRDSIMNYFRTESEGRSPNHIVVLCRGRAFVFDVIHEGCLVTPPELLRQLTYIHKKCHSEPDGPGIAALTSEERTRWAKAREYLIGLDPENLALLEKIQSSLLVYSMEDSSPHVTPEDYSEIIAAILIGDPTVRWGDKSYNLISFSNGVFGCNCDHAPFDAMIMVNISYYVDEKIFQNEGRWKGSEKVRDIPLPEELIFIVDEKVLNDINQAKAQYLREASDLQIAAYAFTSFGKKLTKNKMLHPDTFIQLALQLAYYRLHGHPGCCYETAMTRHFYHGRTETMRSCTVEAVRWCQSMQDPSVNLRERQQKMLQAFAKHNKMMKDCSAGKGFDRHLLGLLLIAKEEGLPVPELFTDPLFSKSGGGGNFVLSTSLVGYLRVQGVVVPMVHNGYGFFYHIRDDRFVVACSAWKSCPETDAEKLVQLTFCAFHDMIQLMNSTHL
RUO
CROT
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
96.6kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
COT
CROT
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit