Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CRSP3 Partial ORF (NP_004821, 531 a.a. - 630 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00009439Q01S

 View more versions of this product

Catalog No. 89-940-578


Add to cart

Description

Description

The activation of gene transcription is a multistep process that is triggered by factors that recognize transcriptional enhancer sites in DNA. These factors work with co-activators to direct transcriptional initiation by the RNA polymerase II apparatus. The protein encoded by this gene is a subunit of the CRSP (cofactor required for SP1 activation) complex, which, along with TFIID, is required for efficient activation by SP1. This protein is also a component of other multisubunit complexes e.g. thyroid hormone receptor-(TR-) associated proteins which interact with TR and facilitate TR function on DNA templates in conjunction with initiation factors and cofactors. This protein also acts as a metastasis suppressor. Two alternatively spliced transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq]

Sequence: LTVHAKMSLIHSIATRVIKLAHAKSSVALAPALVETYSRLLVYMEIESLGIKGFISQLLPTVFKSHAWGILHTLLEMFSYRMHHIQPHYRVQLLSHLHTL
Specifications

Specifications

NP_004821
Liquid
9439
CRSP3 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CRSP130|CRSP133|CRSP3|DKFZp434H0117|DRIP130|SUR2
MED23
Wheat Germ (in vitro)
GST
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
LTVHAKMSLIHSIATRVIKLAHAKSSVALAPALVETYSRLLVYMEIESLGIKGFISQLLPTVFKSHAWGILHTLLEMFSYRMHHIQPHYRVQLLSHLHTL
RUO
MED23
wheat germ expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit