Learn More
Abnova™ Human CRYBA1 Full-length ORF (NP_005199.2, 1 a.a. - 215 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$499.00 - $769.00
Specifications
Accession Number | NP_005199.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 1411 |
Molecular Weight (g/mol) | 51.5kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-940-621
|
Abnova™
H00001411P01L |
25 ug |
Each for $769.00
|
|
|||||
89-940-620
|
Abnova™
H00001411P01S |
10 ug |
Each for $499.00
|
|
|||||
Description
Crystallins are separated into two classes: taxon-specific, or enzyme, and ubiquitous. The latter class constitutes the major proteins of vertebrate eye lens and maintains the transparency and refractive index of the lens. Since lens central fiber cells lose their nuclei during development, these crystallins are made and then retained throughout life, making them extremely stable proteins. Mammalian lens crystallins are divided into alpha, beta, and gamma families; beta and gamma crystallins are also considered as a superfamily. Alpha and beta families are further divided into acidic and basic groups. Seven protein regions exist in crystallins: four homologous motifs, a connecting peptide, and N- and C-terminal extensions. Beta-crystallins, the most heterogeneous, differ by the presence of the C-terminal extension (present in the basic group, none in the acidic group). Beta-crystallins form aggregates of different sizes and are able to self-associate to form dimers or to form heterodimers with other beta-crystallins. This gene, a beta acidic group member, encodes two proteins (crystallin, beta A3 and crystallin, beta A1) from a single mRNA, the latter protein is 17 aa shorter than crystallin, beta A3 and is generated by use of an alternate translation initiation site. Deletion of exons 3 and 4 causes the autosomal dominant disease 'zonular cataract with sutural opacities'. [provided by RefSeq]
Sequence: METQAEQQELETLPTTKMAQTNPTPGSLGPWKITIYDQENFQGKRMEFTSSCPNVSERSFDNVRSLKVESGAWIGYEHTSFCGQQFILERGEYPRWDAWSGSNAYHIERLMSFRPICSANHKESKMTIFEKENFIGRQWEISDDYPSLQAMGWFNNEVGSMKIQSGAWVCYQYPGYRGYQYILECDHHGGDYKHWREWGSHAQTSQIQSIRRIQQSpecifications
NP_005199.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
51.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
METQAEQQELETLPTTKMAQTNPTPGSLGPWKITIYDQENFQGKRMEFTSSCPNVSERSFDNVRSLKVESGAWIGYEHTSFCGQQFILERGEYPRWDAWSGSNAYHIERLMSFRPICSANHKESKMTIFEKENFIGRQWEISDDYPSLQAMGWFNNEVGSMKIQSGAWVCYQYPGYRGYQYILECDHHGGDYKHWREWGSHAQTSQIQSIRRIQQ | |
RUO | |
CRYBA1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
1411 | |
CRYBA1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CRYB1 | |
CRYBA1 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.