Learn More
Abnova™ Human CSF1R (NM_005211.2, 543 a.a. - 972 a.a.) Partial Recombinant Protein with GST-His tag expressed in Sf9 cells
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P4672
Catalog No. 89-940-694
Add to cart
Description
Description
The protein encoded by this gene is the receptor for colony stimulating factor 1, a cytokine which controls the production, differentiation, and function of macrophages. This receptor mediates most if not all of the biological effects of this cytokine. Ligand binding activates the receptor kinase through a process of oligomerization and transphosphorylation. The encoded protein is a tyrosine kinase transmembrane receptor and member of the CSF1/PDGF receptor family of tyrosine-protein kinases. Mutations in this gene have been associated with a predisposition to myeloid malignancy. The first intron of this gene contains a transcriptionally inactive ribosomal protein L7 processed pseudogene oriented in the opposite direction. [provided by RefSeq]
Specifications
Specifications
NM_005211.2
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
77.733kDa
Insect cell (Sf9) expression system
2 ug/lane SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing
C-FMS|CD115|CSFR|FIM2|FMS
CSF1R
50pmol/μg x min
Yes
0.117 ug/uL
Liquid
1436
CSF1R (Human) Recombinant Protein
GST affinity chromatography
100 ug
RUO
CSF1R
Insect cell (Sf9) expression system
Insect
GST-His