Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CSF2 (NP_000749, 18 a.a. - 144 a.a.) Partial Recombinant Protein

Used for Func, SDS-PAGE

Manufacturer:  Abnova™ P3450

Catalog No. 89-940-696


Add to cart

Description

Description

The protein encoded by this gene is a cytokine that controls the production, differentiation, and function of granulocytes and macrophages. The active form of the protein is found extracellularly as a homodimer. This gene has been localized to a cluster of related genes at chromosome region 5q31, which is known to be associated with interstitial deletions in the 5q- syndrome and acute myelogenous leukemia. Other genes in the cluster include those encoding interleukins 4, 5, and 13. [provided by RefSeq]

Sequence: MAPARSPSPSTQPWEHVNAIQEARRLLNLSRDTAAEMNETVEVISEMFDLQEPTCLQTRLELYKQGLRGSLTKLKGPLTMMASHYKQHCPPTPETSCATQIITFESFKENLKDFLLVIPFDCWEPVQE
Specifications

Specifications

NP_000749
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
14.6kDa
Escherichia coli expression system
>95% by SDS-PAGE
100 ug
Store at 4°C for 1-2 weeks. For long term storage store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing.
<1.0 EU/μg of protein (determined by LAL method)
CSF2
Escherichia coli expression system
Escherichia coli
None
1 mg/mL
Liquid
1437
CSF2 (Human) Recombinant Protein
Conventional Chromatography
Loading 3 ug protein in 15% SDS-PAGE
MAPARSPSPSTQPWEHVNAIQEARRLLNLSRDTAAEMNETVEVISEMFDLQEPTCLQTRLELYKQGLRGSLTKLKGPLTMMASHYKQHCPPTPETSCATQIITFESFKENLKDFLLVIPFDCWEPVQE
RUO
GMCSF|MGC131935|MGC138897
CSF2
The ED50 for this effect is < 0.18ng/mL. Measured in a cell proliferation assay using TF1 human erythroleukemic cells.
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit