Learn More
Abnova™ Human CSF2 (P04141, 18 a.a. - 144 a.a.) Full-length Recombinant Protein with N-terminal His tag
Used for Func, SDS-PAGE, WB-Re
Manufacturer:  Abnova™ P5911
Catalog No. 89-940-700
Add to cart
Description
Description
The protein encoded by this gene is a cytokine that controls the production, differentiation, and function of granulocytes and macrophages. The active form of the protein is found extracellularly as a homodimer. This gene has been localized to a cluster of related genes at chromosome region 5q31, which is known to be associated with interstitial deletions in the 5q- syndrome and acute myelogenous leukemia. Other genes in the cluster include those encoding interleukins 4, 5, and 13. [provided by RefSeq]
Sequence: HHHHHHHHHHAPARSPSPSTQPWEHVNAIQEARRLLNLSRDTAAEMNETVEVISEMFDLQEPTCLQTRLELYKQGLRGSLTKLKGPLTMMASHYKQHCPPTPETSCATQIITFESFKENLKDFLLVIPFDCWEPVQE
Specifications
Specifications
P04141
Lyophilized
1437
Non-transgenic plants (Nicotiana benthamiana) expression system
>97% by SDS-PAGE
10 ug
Store at -20°C on dry atmosphere. After reconstitution with sterilized water to a concentration of 25-50 ng/ul, store at -20°C. Aliquot to avoid repeated freezing and thawing.
<0.04 EU/μg protein
CSF2
Non-transgenic plants (Nicotiana benthamiana) expression system
Human ;Human
Yes
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis, Western Blot (Recombinant protein)
Lyophilized
CSF2 (Human) Recombinant Protein
Sequential chromatography (FPLC)
500 ng/lane in 15% SDS-polyacrylamide gel and stained with Coomassie blue
HHHHHHHHHHAPARSPSPSTQPWEHVNAIQEARRLLNLSRDTAAEMNETVEVISEMFDLQEPTCLQTRLELYKQGLRGSLTKLKGPLTMMASHYKQHCPPTPETSCATQIITFESFKENLKDFLLVIPFDCWEPVQE
RUO
GMCSF|MGC131935|MGC138897
CSF2
The activity of recombinant human GM-CSF is determined by the dose-dependent induction of human TF-1 proliferation cell.
Plants
His