Learn More
Abnova™ Human CSF2RA Full-length ORF (NP_006131.2) Recombinant Protein without tag
Used for AP, Func, Screening
Supplier: Abnova™ H00001438G01
Description
The protein encoded by this gene is the alpha subunit of the heterodimeric receptor for colony stimulating factor 2, a cytokine which controls the production, differentiation, and function of granulocytes and macrophages. The encoded protein is a member of the cytokine family of receptors. This gene is found in the pseudoautosomal region (PAR) of the X and Y chromosomes. Multiple transcript variants encoding different isoforms have been found for this gene, with some of the isoforms being membrane-bound and others being soluble. [provided by RefSeq]
Sequence: MLLLVTSLLLCELPHPAFLLIPEKSDLRTVAPASSLNVRFDSRTMNLSWDCQENTTFSKCFLTDKKNRVVEPRLSNNECSCTFREICLHEGVTFEVHVNTSQRGFQQKLLYPNSGREGTAAQNFSCFIYNADLMNCTWARGPTAPRDVQYFLYIRNSKRRREIRCPYYIQDSGTHVGCHLDNLSGLTSRNYFLVNGTSREIGIQFFDSLLDTKKIERFNPPSNVTVRCNTTHCLVRWKQPRTYQKLSYLDFQYQLDVHRKNTQPGTENLLINVSGDLENRYNFPSSEPRAKHSVKIRAADVRILNWSSWSEAIEFGSDDGNLGSVYIYVLLIVGTLVCGIVLGFLFKRFLRIQRLFPPVPQIKDKLNDNHEVEDEIIWEEFTPEEGKGYREEVLTVKEITSpecifications
NP_006131.2 | |
Liquid | |
46.2kDa | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CD116/CDw116/CSF2R/CSF2RAX/CSF2RAY/CSF2RX/CSF2RY/GM-CSF-R-alpha/GMCSFR/GMR/MGC3848/MGC4838 | |
CSF2RA | |
Recombinant | |
wheat germ expression system with proprietary liposome technology |
Antibody Production, Compound Screening, Functional Study | |
1438 | |
CSF2RA (Human) Recombinant Protein | |
MLLLVTSLLLCELPHPAFLLIPEKSDLRTVAPASSLNVRFDSRTMNLSWDCQENTTFSKCFLTDKKNRVVEPRLSNNECSCTFREICLHEGVTFEVHVNTSQRGFQQKLLYPNSGREGTAAQNFSCFIYNADLMNCTWARGPTAPRDVQYFLYIRNSKRRREIRCPYYIQDSGTHVGCHLDNLSGLTSRNYFLVNGTSREIGIQFFDSLLDTKKIERFNPPSNVTVRCNTTHCLVRWKQPRTYQKLSYLDFQYQLDVHRKNTQPGTENLLINVSGDLENRYNFPSSEPRAKHSVKIRAADVRILNWSSWSEAIEFGSDDGNLGSVYIYVLLIVGTLVCGIVLGFLFKRFLRIQRLFPPVPQIKDKLNDNHEVEDEIIWEEFTPEEGKGYREEVLTVKEIT | |
RUO | |
CSF2RA | |
Wheat Germ (in vitro) | |
None | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.