Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CSF3 (190 a.a.) Full-lenth Recombinant Protein with His tag expressed in Barley grain (Hordeum vulgare)

Used for Func, SDS-PAGE, WB-Re

Manufacturer:  Abnova™ P5883

Catalog No. 89-940-712


Add to cart

Description

Description

The protein encoded by this gene is a cytokine that controls the production, differentiation, and function of granulocytes. The active protein is found extracellularly. Three transcript variants encoding three different isoforms have been found for this gene. [provided by RefSeq]

Specifications

Specifications

Func, SDS-PAGE, WB-Re
Lyophilized from PBS, pH 7.2
10.8kDa
Chromatography
SDS-PAGE Stained with Coomassie Blue
Store at −20°C.Aliquot to avoid repeated freezing and thawing.
<0.005ng/μg (0.05EU/μg) as measured by turbidimetric kineticassay.
CSF3
Hordeum vulgare (barley) expression system
Plants
His Tag
Lyophilized
1440
Human CSF3 (190 a.a.) Full-lenth Recombinant Protein with His tag expressed in Barley grain (Hordeum vulgare)
>98% by SDS-PAGE
1mg
RUO
G-CSF, GCSF, MGC45931
CSF3
Activity has been measured by cell-based proliferation assay using mouse NFS-60 cells. The ED50 is typically < 0.1ng/mL.
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit