Learn More
Abnova™ Human CSK (NM_004383, 1 a.a. - 450 a.a.) Full-length Recombinant Protein with GST-His tag expressed in Sf9 cells
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P4673
Catalog No. 89-940-731
Add to cart
Specifications
Specifications
NM_004383
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
80.215kDa
Insect cell (Sf9) expression system
2 ug/lane SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing
MGC117393
CSK
83pmol/μg x min
Yes
0.722 ug/uL
Liquid
1445
CSK (Human) Recombinant Protein
One-step affinity purification using GSH-agarose
100 ug
RUO
CSK
Insect cell (Sf9) expression system
Insect
GST-His