Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CSK (NM_004383) Full-length Recombinant Protein expressed in Sf9 cells

Used for Func, SDS-PAGE

Manufacturer:  Abnova™ P3855

Catalog No. 89-940-729


Add to cart

Specifications

Specifications

NM_004383
Liquid
1445
CSK (Human) Recombinant Protein
>85% by densitometry
5 ug
RUO
CSK
Insect cell (Sf9) expression system
Insect
None
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
78kDa
Insect cell (Sf9) expression system
SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC117393
CSK
The specific activity was determined to be 425nmol/min/mg as per Activity Assay Protocol.
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit