Learn More
Abnova™ Human CSNK1A1 (DQ082865, 1 a.a. - 365 a.a.) Full-length Recombinant Protein with GST-His tag expressed in Sf9 cells
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P4666
Catalog No. 89-940-747
Add to cart
Specifications
Specifications
DQ082865
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
71.434kDa
Insect cell (Sf9) expression system
2 ug/lane SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing
CK1|HLCDGP1|PRO2975
CSNK1A1
3pmol/μg x min
Yes
0.058 ug/uL
Liquid
1452
CSNK1A1 (Human) Recombinant Protein
One-step affinity purification using GSH-agarose
100 ug
RUO
CSNK1A1
Insect cell (Sf9) expression system
Insect
GST-His