Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CSNK1A1 (NP_001883.4, 1 a.a. - 337 a.a.) Full-length Recombinant Protein with GST-tag expressed in Baculovirus infected Sf21 cells

Used for Func, SDS-PAGE

Manufacturer:  Abnova™ P5692

Catalog No. 89-940-748


Add to cart

Specifications

Specifications

NP_001883.4
Liquid
1452
CSNK1A1 (Human) Recombinant Protein
Glutathione sepharose chromatography and anion exchange chromatography
Loading 1 ug protein in SDS-PAGE
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CK1|HLCDGP1|PRO2975
CSNK1A1
The activity was measured by off-chip mobility shift assay. The enzyme was incubated with fluorescence-labeled substrate and Mg (or Mn)/ATP. The phosphorylated and unphosphorylated substrates were separated and detected by LabChip™3000.
Substrate : CHKtide. ATP: 100 µM.
Yes
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
66kDa
Baculovirus expression system
78% by SDS-PAGE/CBB staining.
5 ug
RUO
CSNK1A1
Baculovirus expression system
Insect
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit