Learn More
Abnova™ Human CSNK1E (NP_001885.1, 1 a.a. - 348 a.a.) Partial Recombinant Protein with GST-tag expressed in baculovirus infected Sf21 cells
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P5525
Catalog No. 89-940-756
Add to cart
Description
Description
The protein encoded by this gene is a serine/threonine protein kinase and a member of the casein kinase I protein family, whose members have been implicated in the control of cytoplasmic and nuclear processes, including DNA replication and repair. The encoded protein is found in the cytoplasm as a monomer and can phosphorylate a variety of proteins, including itself. This protein has been shown to phosphorylate period, a circadian rhythm protein. Two transcript variants encoding the same protein have been found for this gene. [provided by RefSeq]
Specifications
Specifications
NP_001885.1
Liquid
1454
CSNK1E (Human) Recombinant Protein
Glutathione sepharose chromatography
Loading 1 ug protein in SDS-PAGE
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HCKIE|MGC10398
CSNK1E
The activity was measured by off-chip mobility shift assay. The enzyme was incubated with fluorescence-labeled substrate and Mg(or Mn)/ATP. The phosphorylated and unphosphorylated substrates were separated and detected by LabChip 3000. Substrate: CKtide. ATP: 100 uM.
Yes
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
68kDa
Baculovirus infected insect cell (Sf21) expression system
54% by SDS-PAGE/CBB staining
5 ug
RUO
CSNK1E
Baculovirus infected insect cell (Sf21) expression system
Insect
GST