Learn More
Abnova™ Human CSNK1G1 (NP_071331.2, 1 a.a. - 422 a.a.) Full-length Recombinant Protein with GST-tag expressed in baculovirus infected Sf21 cells
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P5521
Catalog No. 89-940-760
Add to cart
Description
Description
Casein kinase I is the most abundant serine/threonine kinase in eukaryotic cell extracts. Multiple isoforms of the enzyme exist. The gamma-1 isoform is involved in growth and morphogenesis of eukaryotic cells.[supplied by OMIM]
Specifications
Specifications
NP_071331.2
Liquid
53944
CSNK1G1 (Human) Recombinant Protein
Glutathione sepharose chromatography
Loading 1 ug protein in SDS-PAGE
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CSNK1G1
Baculovirus infected insect cell (Sf21) expression system
Insect
GST
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
76kDa
Baculovirus infected insect cell (Sf21) expression system
95% by SDS-PAGE/CBB staining
5 ug
RUO
CSNK1G1
The activity was measured by off-chip mobility shift assay. The enzyme was incubated with fluorescence-labeled substrate and Mg(or Mn)/ATP. The phosphorylated and unphosphorylated substrates were separated and detected by LabChip 3000. Substrate: CKtide. ATP: 100 uM.
Yes