Learn More
Abnova™ Human CSNK1G2 (NP_001310.3, 1 a.a. - 415 a.a.) Full-length Recombinant Protein with GST-tag expressed in baculovirus infected Sf21 cells
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P5522
Catalog No. 89-940-763
Add to cart
Specifications
Specifications
NP_001310.3
Liquid
1455
CSNK1G2 (Human) Recombinant Protein
Glutathione sepharose chromatography
Loading 1 ug protein in SDS-PAGE
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CK1g2
CSNK1G2
The activity was measured by off-chip mobility shift assay. The enzyme was incubated with fluorescence-labeled substrate and Mg(or Mn)/ATP. The phosphorylated and unphosphorylated substrates were separated and detected by LabChip 3000. Substrate: CKtide. ATP: 100 uM.
Yes
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
74kDa
Baculovirus infected insect cell (Sf21) expression system
95% by SDS-PAGE/CBB staining
5 ug
RUO
CSNK1G2
Baculovirus infected insect cell (Sf21) expression system
Insect
GST