Learn More
Abnova™ Human CSNK1G3 (NP_004375.2, 1 a.a. - 447 a.a.) Full-length Recombinant Protein with GST-tag expressed in baculovirus infected Sf21 cells
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P5523
Catalog No. 89-940-766
Add to cart
Description
Description
Specifications
Specifications
Baculovirus infected insect cell (Sf21) expression system
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
78kDa
Baculovirus infected insect cell (Sf21) expression system
94% by SDS-PAGE/CBB staining
5 ug
RUO
CSNK1G3
Insect
GST
NP_004375.2
Liquid
1456
CSNK1G3 (Human) Recombinant Protein
Glutathione sepharose chromatography
Loading 1 ug protein in SDS-PAGE
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CSNK1G3
The activity was measured by off-chip mobility shift assay. The enzyme was incubated with fluorescence-labeled substrate and Mg(or Mn)/ATP. The phosphorylated and unphosphorylated substrates were separated and detected by LabChip 3000. Substrate: CKtide. ATP: 100 uM.
Yes