Learn More
Abnova™ Human CSNK2A1 (NM_001895) Full-length Recombinant Protein with GST expressed in Sf9 cells
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P4221
Catalog No. 89-940-770
Add to cart
Description
Description
Casein kinase II is a serine/threonine protein kinase that phosphorylates acidic proteins such as casein. The kinase exists as a tetramer and is composed of an alpha, an alpha-prime, and two beta subunits. The alpha subunits contain the catalytic activity while the beta subunits undergo autophosphorylation. The protein encoded by this gene represents the alpha subunit. While this gene is found on chromosome 20, a related transcribed pseudogene is found on chromosome 11. Three transcript variants encoding two different proteins have been found for this gene. [provided by RefSeq]
Specifications
Specifications
NM_001895
Liquid
1457
CSNK2A1 (Human) Recombinant Protein
2 ug/lane SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CK2A1|CKII
CSNK2A1
Insect
GST
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
70kDa
≥90% by SDS-PAGE
5 ug
RUO
CSNK2A1
107nmol phosphate incorporated into casein per minute per mg protein at 30°C for 15 minutes using a final concentration of 50μM ATP (0.83 uCi/assay).
Yes