Learn More
Abnova™ Human CSNK2A2 (NM_001896.2, 1 a.a. - 350 a.a.) Full-length Recombinant Protein with GST-His tag expressed in Sf9 cells
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P4668
Catalog No. 89-940-778
Add to cart
Specifications
Specifications
NM_001896.2
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
71.109kDa
Insect cell (Sf9) expression system
2 ug/lane SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing
CK2A2|CSNK2A1|FLJ43934
CSNK2A2
2pmol/μg x min
Yes
0.715 ug/uL
Liquid
1459
CSNK2A2 (Human) Recombinant Protein
One-step affinity purification using GSH-agarose
100 ug
RUO
CSNK2A2
Insect cell (Sf9) expression system
Insect
GST-His