Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CSRP2BP Full-length ORF (NP_065397.1, 1 a.a. - 782 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00057325P01

Catalog No. 89-940-794


Add to cart

Description

Description

CSRP2 is a protein containing two LIM domains, which are double zinc finger motifs found in proteins of diverse function. CSRP2 and some related proteins are thought to act as protein adapters, bridging two or more proteins to form a larger protein complex. The protein encoded by this gene binds to one of the LIM domains of CSRP2 and contains an acetyltransferase domain. Although the encoded protein has been detected in the cytoplasm, it is predominantly a nuclear protein. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MDSSIHLSSLISRHDDEATRTSTSEGLEEGEVEGETLLIVESEDQASVDLSHDQSGDSLNSDEGDVSWMEEQLSYFCDKCQKWIPASQLREQLSYLKGDNFFRFTCSDCSADGKEQYERLKLTWQQVVMLAMYNLSLEGSGRQGYFRWKEDICAFIEKHWTFLLGNRKKTSTWWSTVAGCLSVGSPMYFRSGAQEFGEPGWWKLVHNKPPTMKPEGEKLSASTLKIKAASKPTLDPIITVEGLRKRASRNPVESAMELKEKRSRTQEAKDIRRAQKEAAGFLDRSTSSTPVKFISRGRRPDVILEKGEVIDFSSLSSSDRTPLTSPSPSPSLDFSAPGTPASHSATPSLLSEADLIPDVMPPQALFHDDDEMEGDGVIDPGMEYVPPPAGSVASGPVVGGRKKVRGPEQIKQEVESEEEKPDRMDIDSEDTDSNTSLQTRAREKRKPQLEKDTKPKEPRYTPVSIYEEKLLLKRLEACPGAVAMTPEARRLKRKLIVRQAKRDRGLPLFDLDQVVNAALLLVDGIYGAKEGGISRLPAGQATYRTTCQDFRILDRYQTSLPSRKGFRHQTTKFLYRLVGSEDMAVDQSIVSPYTSRILKPYIRRDYETKPPKLQLLSQIRSHLHRSDPHWTPEPDAPLDYCYVRPNHIPTINSMCQEFFWPGIDLSECLQYPDFSVVVLYKKVIIAFGFMVPDVKYNEAYISFLFVHPEWRRAGIATFMIYHLIQTCMGKDVTLHVSASNPAMLLYQKFGFKTEEYVLDFYDKYYPLESTECKHAFFLRLRR
Specifications

Specifications

NP_065397.1
Liquid
57325
CSRP2BP (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MDSSIHLSSLISRHDDEATRTSTSEGLEEGEVEGETLLIVESEDQASVDLSHDQSGDSLNSDEGDVSWMEEQLSYFCDKCQKWIPASQLREQLSYLKGDNFFRFTCSDCSADGKEQYERLKLTWQQVVMLAMYNLSLEGSGRQGYFRWKEDICAFIEKHWTFLLGNRKKTSTWWSTVAGCLSVGSPMYFRSGAQEFGEPGWWKLVHNKPPTMKPEGEKLSASTLKIKAASKPTLDPIITVEGLRKRASRNPVESAMELKEKRSRTQEAKDIRRAQKEAAGFLDRSTSSTPVKFISRGRRPDVILEKGEVIDFSSLSSSDRTPLTSPSPSPSLDFSAPGTPASHSATPSLLSEADLIPDVMPPQALFHDDDEMEGDGVIDPGMEYVPPPAGSVASGPVVGGRKKVRGPEQIKQEVESEEEKPDRMDIDSEDTDSNTSLQTRAREKRKPQLEKDTKPKEPRYTPVSIYEEKLLLKRLEACPGAVAMTPEARRLKRKLIVRQAKRDRGLPLFDLDQVVNAALLLVDGIYGAKEGGISRLPAGQATYRTTCQDFRILDRYQTSLPSRKGFRHQTTKFLYRLVGSEDMAVDQSIVSPYTSRILKPYIRRDYETKPPKLQLLSQIRSHLHRSDPHWTPEPDAPLDYCYVRPNHIPTINSMCQEFFWPGIDLSECLQYPDFSVVVLYKKVIIAFGFMVPDVKYNEAYISFLFVHPEWRRAGIATFMIYHLIQTCMGKDVTLHVSASNPAMLLYQKFGFKTEEYVLDFYDKYYPLESTECKHAFFLRLRR
RUO
CSRP2BP
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
115.2kDa
Glutathione Sepharose 4 Fast Flow
2 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CRP2BP|MGC15388|PRO1194|dJ717M23.1
CSRP2BP
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit