Learn More
Abnova™ Human CTGF (P29279) Recombinant Protein
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P4578
Catalog No. 89-940-872
Add to cart
Description
Description
Sequence: MGKKCIRTPKISKPIKFELSGCTSMKTYRAKFCGVCTDGRCCTPHRTTTLPVEFKCPDGEVMKKNMMFIKTCACHYNCPGDNDIFESLYYRKMYGDMA
Specifications
Specifications
P29279
Lyophilized
1490
CTGF (Human) Recombinant Protein
1 ug/lane in 4-20% Tris-Glycine gel Stained with Coomassie Blue
MGKKCIRTPKISKPIKFELSGCTSMKTYRAKFCGVCTDGRCCTPHRTTTLPVEFKCPDGEVMKKNMMFIKTCACHYNCPGDNDIFESLYYRKMYGDMA
RUO
CCN2|HCS24|IGFBP8|MGC102839|NOV2
CTGF
The activity is determined by the dose-dependent induction of HUVEC proliferation. The expected ED50 for this effect is 1.1-1.7μg/mL.
Yes
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Lyophilized
11.2kDa
Escherichia coli expression system
20 ug
Store at -20°C on dry atmosphere. After reconstitution with 5 mM sodium acetate, store at -20°C. Aliquot to avoid repeated freezing and thawing.
<0.1 EU/μg
CTGF
Escherichia coli expression system
Escherichia coli
None