Learn More
Abnova™ Human CTLA4 Recombinant Protein fused to murine IgG2a Fc purified from CHO cells
Used for SDS-PAGE
Manufacturer:  Abnova™ P4933
Catalog No. 89-940-882
Add to cart
Description
Description
This gene is a member of the immunoglobulin superfamily and encodes a protein which transmits an inhibitory signal to T cells. The protein contains a V domain, a transmembrane domain, and a cytoplasmic tail. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. The membrane-bound isoform functions as a homodimer interconnected by a disulfide bond, while the soluble isoform functions as a monomer. Mutations in this gene have been associated with insulin-dependent diabetes mellitus, Graves disease, Hashimoto thyroiditis, celiac disease, systemic lupus erythematosus, thyroid-associated orbitopathy, and other autoimmune diseases. [provided by RefSeq]
Sequence: CTLA4 mature: mhvaqpavvlassrgiasfvceyaspgkatevrvtvlrqadsqvtevcaatymtgneltflddsictgtssgnqvnltiqglramdtglyickvelmypppyylgigngtqiyvidpepcpdsdf