Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CTNS Full-length ORF (AAH32850.1, 1 a.a. - 400 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00001497P01

Catalog No. 89-940-914


Add to cart

Description

Description

This gene encodes a seven-transmembrane domain protein that functions to transport cystine out of lysosomes. Its activity is driven by the H+ electrochemical gradient of the lysosomal membrane. Mutations in this gene cause cystinosis, a lysosomal storage disorder. Alternative splicing results in multiple transcript variants. [provided by RefSeq]

Sequence: MIRNWLTIFILFPLKLVEKCESSVSLTVPPVVKLENGSSTNVSLTLRPPLNATLVITFEITFRSKNITILELPDEVVVPPGVTNSSFQVTSQNVGQLTVYLHGNHSNQTGPRIRFLVIRSSAISIINQVIGWIYFVAWSISFYPQVIMNWRRKSVIGLSFDFVALNLTGFVAYSVFNIGLLWVPYIKEQFLLKYPNGVNPVNSNDVFFSLHAVVLTLIIIVQCCLYERGGQRVSWPAIGFLVLAWLFAFVTMIVAAVGVITWLQFLFCFSYIKLAVTLVKYFPQAYMNFYYKSTEGWSIGNVLLDFTGGSFSLLQMFLQSYNNDQWTLIFGDPTKFGLGVFSIVFDVVFFIQHFCLYRKRPGLQAARTGSGSRLRQDWAPSLQPKALPQTTSVSASSLKG
Specifications

Specifications

AAH32850.1
Liquid
1497
CTNS (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MIRNWLTIFILFPLKLVEKCESSVSLTVPPVVKLENGSSTNVSLTLRPPLNATLVITFEITFRSKNITILELPDEVVVPPGVTNSSFQVTSQNVGQLTVYLHGNHSNQTGPRIRFLVIRSSAISIINQVIGWIYFVAWSISFYPQVIMNWRRKSVIGLSFDFVALNLTGFVAYSVFNIGLLWVPYIKEQFLLKYPNGVNPVNSNDVFFSLHAVVLTLIIIVQCCLYERGGQRVSWPAIGFLVLAWLFAFVTMIVAAVGVITWLQFLFCFSYIKLAVTLVKYFPQAYMNFYYKSTEGWSIGNVLLDFTGGSFSLLQMFLQSYNNDQWTLIFGDPTKFGLGVFSIVFDVVFFIQHFCLYRKRPGLQAARTGSGSRLRQDWAPSLQPKALPQTTSVSASSLKG
RUO
CTNS
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
71.5kDa
Glutathione Sepharose 4 Fast Flow
2 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CTNS-LSB|PQLC4
CTNS
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit