Learn More
Novus Biologicals™ Human CTRP2 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer: Novus Biologicals™ NBP266752
Description
The Human CTRP2 ELISA Kit (Colorimetric) measures human CTRP2 in serum, plasma and other biological fluids.Specifications
0.31 - 20 ng/mL | |
This kit recognizes Human C1QTNF2 in samples. No significant cross-reactivity or interference between Human C1QTNF2 and analogues was observed. | |
114898 | |
Human | |
1 Kit | |
Storage is content dependent. |
0.19 ng/mL | |
ELISA | |
C1QTNF2 | |
ELISA Kit (Colorimetric) | |
Biological Fluids,Plasma,Serum | |
CTRP2 |