Learn More
Novus Biologicals™ Human CTRP9/C1qTNF9 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP266754
Catalog No. NBP266754
Add to cart
Description
Description
The Human CTRP9/C1qTNF9 ELISA Kit (Colorimetric) measures human CTRP9/C1qTNF9 in serum, plasma and other biological fluids.
Specifications
Specifications
1 Kit
0.16 - 10 ng/mL
ELISA
ELISA Kit (Colorimetric)
Storage is content dependent.
Human
Biological Fluids, Plasma, Serum
This kit recognizes Human C1QTNF9 in samples. No significant cross-reactivity or interference between Human C1QTNF9 and analogues was observed.
338872
0.10 ng/mL
CTRP9/C1qTNF9
C1QTNF9