Learn More
Abnova™ Human CTSD (P07339) native Protein from human liver
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P4030
Catalog No. 89-940-938
Add to cart
Description
Description
This gene encodes a lysosomal aspartyl protease composed of a dimer of disulfide-linked heavy and light chains, both produced from a single protein precursor. This proteinase, which is a member of the peptidase C1 family, has a specificity similar to but narrower than that of pepsin A. Transcription of this gene is initiated from several sites, including one which is a start site for an estrogen-regulated transcript. Mutations in this gene are involved in the pathogenesis of several diseases, including breast cancer and possibly Alzheimer disease. [provided by RefSeq]